Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTCTGTACTCTGGTGGCTTCCA[A/G]TGGAAAAGGCATTTTGGAGGCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172470 MIM: 607940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHKG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHKG1 - phosphorylase kinase catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042469.1 | 449 | Intron | NP_001035934.2 | |||
NM_001042470.1 | 449 | Intron | NP_001035935.2 | |||
NM_001130069.2 | 449 | Missense Mutation | ATG,GTG | M,V 140 | NP_001123541.1 | |
NM_001146333.1 | 449 | Missense Mutation | ATG,GTG | M,V 33 | NP_001139805.1 | |
NM_015411.2 | 449 | Missense Mutation | ATG,GTG | M,V 140 | NP_056226.2 | |
XM_011515254.2 | 449 | Missense Mutation | ATG,GTG | M,V 140 | XP_011513556.1 | |
XM_017011939.1 | 449 | Missense Mutation | ATG,GTG | M,V 140 | XP_016867428.1 | |
XM_017011940.1 | 449 | Intron | XP_016867429.1 |