Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGGTGTCTAAAGAGGTCAGAG[C/T]GTTGTTTGAAACTCTTCCCACAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZKSCAN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZKSCAN5 - zinc finger with KRAB and SCAN domains 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF394 - zinc finger protein 394 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032164.2 | 1191 | Missense Mutation | NP_115540.2 | |||
XM_017012710.1 | 1191 | Intron | XP_016868199.1 | |||
XM_017012711.1 | 1191 | UTR 3 | XP_016868200.1 |
ZNF789 - zinc finger protein 789 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |