Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGACGAGGGAGGATTGTTCGG[G/T]TCATACACGAAGAGCTTCTGGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602447 MIM: 602720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PON2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PON2 - paraoxonase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000305.2 | 878 | Missense Mutation | GAA,GAC | E,D 294 | NP_000296.2 | |
NM_001018161.1 | 878 | Missense Mutation | GAA,GAC | E,D 282 | NP_001018171.1 | |
XM_005250453.1 | 878 | Missense Mutation | GAA,GAC | E,D 226 | XP_005250510.1 | |
XM_017012357.1 | 878 | Missense Mutation | GAA,GAC | E,D 224 | XP_016867846.1 | |
XM_017012358.1 | 878 | Missense Mutation | GAA,GAC | E,D 208 | XP_016867847.1 |
PON3 - paraoxonase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |