Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACGGCTGGGGCAGTGGGTCCC[C/T]GCCCTCAGCGGTGTGCGTTGAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616813 MIM: 606965 MIM: 109280 MIM: 614792 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGAP3 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FASTK - Fas activated serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A2 - solute carrier family 4 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMUB1 - transmembrane and ubiquitin like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136044.1 | 239 | Missense Mutation | AGG,GGG | R,G 37 | NP_001129516.1 | |
NM_031434.3 | 239 | Missense Mutation | AGG,GGG | R,G 37 | NP_113622.1 |