Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACCCATCCTCATCTGGAACTCCA[G/T]TGTTAGGGTCACAGTCCCGGACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604355 MIM: 601029 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPG2 - coatomer protein complex subunit gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290033.1 | 2297 | Intron | NP_001276962.1 | |||
NM_012133.5 | 2297 | Missense Mutation | AAT,ACT | N,T 736 | NP_036265.3 |
MEST - mesoderm specific transcript | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |