Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGAGTTTGTTCCAAAACATGCA[A/C]GACAATGTAATACCCTTGCCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927391 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927391 - uncharacterized LOC101927391 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIOS - meiosis regulator for oocyte development | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019005.3 | 474 | Silent Mutation | AGA,CGA | R,R 114 | NP_061878.3 | |
XM_005249780.3 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_005249837.1 | |
XM_005249781.3 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_005249838.1 | |
XM_005249782.3 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_005249839.1 | |
XM_005249784.3 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_005249841.1 | |
XM_011515432.2 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_011513734.1 | |
XM_017012361.1 | 474 | Silent Mutation | AGA,CGA | R,R 114 | XP_016867850.1 | |
XM_017012362.1 | 474 | UTR 5 | XP_016867851.1 |