Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATCTCCCCGTTTTTCATGATCT[G/T]CTTGATCTTGGATCTTTTGTTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600028 MIM: 600030 | ||||||||||||||||||||
Literature Links: |
DLX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLX5 - distal-less homeobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005221.5 | 1899 | Missense Mutation | AAG,CAG | K,Q 194 | NP_005212.1 | |
XM_005250185.3 | 1899 | Missense Mutation | AAG,CAG | K,Q 66 | XP_005250242.1 | |
XM_017011803.1 | 1899 | Missense Mutation | AAG,CAG | K,Q 66 | XP_016867292.1 |
DLX6 - distal-less homeobox 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DLX6-AS1 - DLX6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |