Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGCAGGCTATAGAGAGGAAGG[G/T]GGAGATCCACAGCCGGCGACTCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF398 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF398 - zinc finger protein 398 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020781.3 | 281 | UTR 5 | NP_065832.1 | |||
NM_170686.2 | 281 | Missense Mutation | GGG,GTG | G,V 57 | NP_733787.1 | |
XM_011516439.2 | 281 | Missense Mutation | GGG,GTG | G,V 62 | XP_011514741.1 | |
XM_011516440.2 | 281 | Missense Mutation | GGG,GTG | G,V 50 | XP_011514742.1 | |
XM_011516441.2 | 281 | UTR 5 | XP_011514743.1 |