Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCACCTCCAGCAGAGCTCGCAT[G/T]ATCTTTCTAGGGTGGGGCACACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172470 MIM: 607940 | ||||||||||||||||||||
Literature Links: |
PHKG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHKG1 - phosphorylase kinase catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258459.1 | 558 | Silent Mutation | NP_001245388.1 | |||
NM_001258460.1 | 558 | Silent Mutation | NP_001245389.1 | |||
NM_006213.4 | 558 | Silent Mutation | NP_006204.1 | |||
XM_005271772.4 | 558 | Silent Mutation | XP_005271829.1 | |||
XM_017012324.1 | 558 | Silent Mutation | XP_016867813.1 | |||
XM_017012325.1 | 558 | Silent Mutation | XP_016867814.1 | |||
XM_017012326.1 | 558 | Silent Mutation | XP_016867815.1 | |||
XM_017012327.1 | 558 | Silent Mutation | XP_016867816.1 |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |