Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAGGCAGAAGCCCTGGCAGGGC[A/G]TAAGTGAGGGGACCCCCCGGCACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608235 MIM: 608489 | ||||||||||||||||||||
Literature Links: |
GAL3ST4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAL3ST4 - galactose-3-O-sulfotransferase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPC2 - glypican 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152742.2 | 1036 | Missense Mutation | ACG,ATG | T,M 270 | NP_689955.1 |
STAG3 - stromal antigen 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |