Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTAGCTGCATCTTGGAACCCGA[A/G]GGGCTGGTGGCCTGCGGGGCAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602686 | ||||||||||||||||||||
Literature Links: |
MAD1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAD1L1 - MAD1 mitotic arrest deficient like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013836.1 | 1935 | Silent Mutation | CCC,CCT | P,P 670 | NP_001013858.1 | |
NM_001013837.1 | 1935 | Silent Mutation | CCC,CCT | P,P 670 | NP_001013859.1 | |
NM_001304523.1 | 1935 | Silent Mutation | CCC,CCT | P,P 670 | NP_001291452.1 | |
NM_001304524.1 | 1935 | Silent Mutation | CCC,CCT | P,P 578 | NP_001291453.1 | |
NM_001304525.1 | 1935 | Silent Mutation | CCC,CCT | P,P 126 | NP_001291454.1 | |
NM_003550.2 | 1935 | Silent Mutation | CCC,CCT | P,P 670 | NP_003541.2 | |
XM_005249877.1 | 1935 | Silent Mutation | CCC,CCT | P,P 623 | XP_005249934.1 | |
XM_011515567.1 | 1935 | Silent Mutation | CCC,CCT | P,P 651 | XP_011513869.1 | |
XM_011515568.2 | 1935 | Intron | XP_011513870.1 | |||
XM_011515569.2 | 1935 | Silent Mutation | CCC,CCT | P,P 348 | XP_011513871.2 | |
XM_011515571.1 | 1935 | Silent Mutation | CCC,CCT | P,P 260 | XP_011513873.1 | |
XM_017012690.1 | 1935 | Silent Mutation | CCC,CCT | P,P 734 | XP_016868179.1 | |
XM_017012691.1 | 1935 | Silent Mutation | CCC,CCT | P,P 260 | XP_016868180.1 |