Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCGGGGGCGCCGGCGAGCCCC[C/G]CAAACCAGACAAGTTATACGGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PRR15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRR15 - proline rich 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175887.2 | 1730 | Missense Mutation | CCC,CGC | P,R 72 | NP_787083.1 | |
XM_005249653.3 | 1730 | Missense Mutation | CCC,CGC | P,R 72 | XP_005249710.1 | |
XM_011515198.1 | 1730 | Missense Mutation | CCC,CGC | P,R 72 | XP_011513500.1 | |
XM_011515199.1 | 1730 | Missense Mutation | CCC,CGC | P,R 72 | XP_011513501.1 | |
XM_017011842.1 | 1730 | Missense Mutation | CCC,CGC | P,R 72 | XP_016867331.1 |