Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAACTGCGCGGACAAGCTGCAGC[A/G]ACTGGGTGAGGGCACGTCTGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610039 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS37D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS37D - VPS37D, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077621.1 | 420 | Missense Mutation | CAA,CGA | Q,R 102 | NP_001071089.1 | |
XM_017011779.1 | 420 | Missense Mutation | CAA,CGA | Q,R 61 | XP_016867268.1 |