Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAACTGCTGGCCCTTCTTGGT[A/G]GTGAAGCTGTGGAAGAAAGGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602495 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL24 - C-C motif chemokine ligand 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002991.2 | 465 | Silent Mutation | ACC,ACT | T,T 66 | NP_002982.2 | |
XM_011516459.2 | 465 | Silent Mutation | ACC,ACT | T,T 66 | XP_011514761.1 | |
XM_011516460.2 | 465 | Silent Mutation | ACC,ACT | T,T 66 | XP_011514762.1 |