Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCTGTCGCCCAGTGTTCCCTTG[C/T]CGTCGGCGATTTTGCCCCCGACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPED1 - cadherin like and PC-esterase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105533.1 | 258 | Silent Mutation | TGC,TGT | C,C 9 | NP_001099003.1 | |
NM_024913.4 | 258 | Silent Mutation | TGC,TGT | C,C 9 | NP_079189.4 | |
XM_017012649.1 | 258 | Silent Mutation | TGC,TGT | C,C 9 | XP_016868138.1 |