Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTTGAGCCGAGCCAACTCAGCC[C/T]TGAGCCTCGCCTGGTGCCGGCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615732 MIM: 612548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSUN5 - NOP2/Sun RNA methyltransferase family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168347.2 | Intron | NP_001161819.1 | ||||
NM_001168348.2 | Intron | NP_001161820.1 | ||||
NM_018044.4 | Intron | NP_060514.1 | ||||
NM_148956.3 | Intron | NP_683759.1 | ||||
XM_005277604.4 | Intron | XP_005277661.1 |
TRIM50 - tripartite motif containing 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |