Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAACTCTACCCAGTGTGGAAGC[A/G]GGGAGACATGAGGTGGAAAAACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604368 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPNMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPNMB - glycoprotein nmb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005340.1 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | NP_001005340.1 | |
NM_002510.2 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | NP_002501.1 | |
XM_005249578.2 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | XP_005249635.1 | |
XM_017011676.1 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | XP_016867165.1 | |
XM_017011677.1 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | XP_016867166.1 | |
XM_017011678.1 | 843 | Missense Mutation | CAG,CGG | Q,R 64 | XP_016867167.1 |