Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCACCGCCGAGACCAGGGCCG[T/C]GCCCCGGCCACTGCCCTCCTCCGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138079 MIM: 613993 | ||||||||||||||||||||
Literature Links: |
GCK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCK - glucokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000162.3 | 1815 | Missense Mutation | ACG,GCG | T,A 449 | NP_000153.1 | |
NM_033507.1 | 1815 | Missense Mutation | ACG,GCG | T,A 450 | NP_277042.1 | |
NM_033508.1 | 1815 | Missense Mutation | ACG,GCG | T,A 448 | NP_277043.1 | |
XM_017011966.1 | 1815 | Missense Mutation | ACG,GCG | T,A 449 | XP_016867455.1 |
MYL7 - myosin light chain 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |