Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCCAGCTCGGGGGAGGACAGAA[A/G]AAAAAGGAGGTAGAATGGATCCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601385 | ||||||||||||||||||||
Literature Links: |
TUSC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUSC3 - tumor suppressor candidate 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006765.3 | 473 | Silent Mutation | AAA,AAG | K,K 43 | NP_006756.2 | |
NM_178234.2 | 473 | Silent Mutation | AAA,AAG | K,K 43 | NP_839952.1 | |
XM_011544651.2 | 473 | Intron | XP_011542953.1 | |||
XM_011544652.2 | 473 | Intron | XP_011542954.1 | |||
XM_011544654.2 | 473 | Intron | XP_011542956.1 | |||
XM_017013860.1 | 473 | Intron | XP_016869349.1 | |||
XM_017013861.1 | 473 | Intron | XP_016869350.1 |