Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615880 MIM: 603780 | ||||||||||||||||||||
Literature Links: |
ARHGAP39 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP39 - Rho GTPase activating protein 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C8orf82 - chromosome 8 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC14 - leucine rich repeat containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272036.1 | 461 | Silent Mutation | GGG,GGT | G,G 153 | NP_001258965.1 | |
NM_014665.3 | 461 | Silent Mutation | GGG,GGT | G,G 153 | NP_055480.1 | |
XM_005272358.4 | 461 | Silent Mutation | GGG,GGT | G,G 153 | XP_005272415.1 | |
XM_005272359.4 | 461 | Silent Mutation | GGG,GGT | G,G 153 | XP_005272416.1 | |
XM_005272360.4 | 461 | Silent Mutation | GGG,GGT | G,G 114 | XP_005272417.1 | |
XM_017014005.1 | 461 | Silent Mutation | GGG,GGT | G,G 114 | XP_016869494.1 |
LRRC24 - leucine rich repeat containing 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD3 - major facilitator superfamily domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RECQL4 - RecQ like helicase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |