Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCTGCCGGTAGTCTCTCTCCC[G/T]CAGCTGGGCGCGCACCTCGAACGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SGK223 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGK223 - homolog of rat pragma of Rnd2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080826.2 | 4507 | Silent Mutation | AGG,CGG | R,R 1278 | NP_001074295.2 | |
XM_005272369.4 | 4507 | Silent Mutation | AGG,CGG | R,R 1278 | XP_005272426.2 | |
XM_005272370.4 | 4507 | Silent Mutation | AGG,CGG | R,R 1278 | XP_005272427.2 | |
XM_011543813.2 | 4507 | Intron | XP_011542115.1 |