Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGTTGAGTCCAAGATAGCCGTG[A/G]GTCCCACTCTGCTCGGTGATGTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GOT1L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOT1L1 - glutamic-oxaloacetic transaminase 1-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152413.2 | 1430 | Silent Mutation | ACC,ACT | T,T 350 | NP_689626.2 | |
XM_005273399.3 | 1430 | Silent Mutation | ACC,ACT | T,T 349 | XP_005273456.1 | |
XM_006716285.3 | 1430 | Silent Mutation | ACC,ACT | T,T 319 | XP_006716348.1 | |
XM_011544391.2 | 1430 | Silent Mutation | ACC,ACT | T,T 168 | XP_011542693.1 |