Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAAGTCCCCGGAGGTATCTGAGC[A/G]AACCATGTCAGAGAGAGACTTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA13 - annexin A13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003954.2 | 990 | Missense Mutation | CGC,TGC | R,C 341 | NP_001003954.1 | |
NM_004306.3 | 990 | Missense Mutation | CGC,TGC | R,C 300 | NP_004297.2 |