Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTGATTTCCTCAAACTCTTCTC[C/T]CAGCTTAAAGGAGATCTCATTATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FABP12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FABP12 - fatty acid binding protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105281.1 | 202 | Nonsense Mutation | NP_001098751.1 | |||
XM_005251285.4 | 202 | Nonsense Mutation | XP_005251342.1 | |||
XM_006716465.3 | 202 | Nonsense Mutation | XP_006716528.1 | |||
XM_011517577.2 | 202 | Nonsense Mutation | XP_011515879.1 |