Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCTGGCAAGGCTGAGAAACAAT[C/G]ATTTCACATCTTGAGTGAAACAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM71 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM71 - transmembrane protein 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145153.1 | 996 | Nonsense Mutation | TCA,TGA | S,* 212 | NP_001138625.1 | |
NM_144649.2 | 996 | Nonsense Mutation | TCA,TGA | S,* 256 | NP_653250.2 |