Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGGTCTGGCTCTGCTGGGGCCT[A/G]AGTTAGGTTTGATCACCTTGTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601956 | ||||||||||||||||||||
Literature Links: |
GFRA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFRA2 - GDNF family receptor alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165038.1 | 1682 | Missense Mutation | TCA,TTA | S,L 336 | NP_001158510.1 | |
NM_001165039.1 | 1682 | Missense Mutation | TCA,TTA | S,L 308 | NP_001158511.1 | |
NM_001495.4 | 1682 | Missense Mutation | TCA,TTA | S,L 441 | NP_001486.4 | |
XM_006716327.3 | 1682 | Missense Mutation | TCA,TTA | S,L 441 | XP_006716390.1 | |
XM_011544484.2 | 1682 | Missense Mutation | TCA,TTA | S,L 441 | XP_011542786.1 |