Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTTCGGGGACGGCAGGAGCAC[A/G]GACATCCAGGAGGAGGCCCTCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
AARD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AARD - alanine and arginine rich domain containing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025357.2 | 148 | Silent Mutation | ACA,ACG | T,T 43 | NP_001020528.1 | |
XM_011517043.2 | 148 | Silent Mutation | ACA,ACG | T,T 43 | XP_011515345.1 |