Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCTCGCGCGGCCCAAGTCTCTA[A/G]TGCAGAAGCTCCAATGCTCCTTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZBTB10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB10 - zinc finger and BTB domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105539.2 | 1325 | Missense Mutation | ATG,GTG | M,V 243 | NP_001099009.1 | |
NM_001277145.1 | 1325 | Intron | NP_001264074.1 | |||
NM_023929.4 | 1325 | Missense Mutation | ATG,GTG | M,V 243 | NP_076418.3 |