Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTCAGCTAAGTGTGTATCTGGA[A/G]CAGGACAGGTGAGCAAGAAACCGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM92A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM92A1 - family with sequence similarity 92 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283034.1 | 10825 | Intron | NP_001269963.1 | |||
NM_145269.4 | 10825 | Intron | NP_660312.2 | |||
XM_005250787.2 | 10825 | Missense Mutation | ACA,GCA | T,A 187 | XP_005250844.1 |
RBM12B - RNA binding motif protein 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203390.2 | 10825 | Intron | NP_976324.2 | |||
XM_005250915.4 | 10825 | Intron | XP_005250972.1 | |||
XM_011517027.2 | 10825 | UTR 3 | XP_011515329.1 | |||
XM_011517028.2 | 10825 | Intron | XP_011515330.1 | |||
XM_011517029.2 | 10825 | Intron | XP_011515331.1 | |||
XM_017013406.1 | 10825 | Intron | XP_016868895.1 | |||
XM_017013407.1 | 10825 | Intron | XP_016868896.1 | |||
XM_017013408.1 | 10825 | Intron | XP_016868897.1 |