Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGTTTTTCTTTATAAGATATG[C/T]GTAACACACCAACGTACTGTGACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605417 MIM: 610029 | ||||||||||||||||||||
Literature Links: |
DKK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DKK4 - dickkopf WNT signaling pathway inhibitor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VDAC3 - voltage dependent anion channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135694.2 | 214 | Missense Mutation | CGT,TGT | R,C 2 | NP_001129166.1 | |
NM_005662.6 | 214 | Missense Mutation | CGT,TGT | R,C 2 | NP_005653.3 | |
XM_006716394.1 | 214 | UTR 5 | XP_006716457.1 |