Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCACAGCCCCCTGATGCTGAAC[A/G]ACAGTGGCTCAGCACATTCCATGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604900 MIM: 140580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGAT1 - diacylglycerol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSF1 - heat shock transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005272315.2 | 813 | Missense Mutation | AAC,GAC | N,D 215 | XP_005272372.1 | |
XM_005272316.2 | 813 | Missense Mutation | AAC,GAC | N,D 215 | XP_005272373.1 | |
XM_005272317.1 | 813 | Missense Mutation | AAC,GAC | N,D 215 | XP_005272374.1 | |
XM_011517004.2 | 813 | Missense Mutation | AAC,GAC | N,D 4 | XP_011515306.1 | |
XM_011517006.2 | 813 | UTR 5 | XP_011515308.1 | |||
XM_017013377.1 | 813 | Missense Mutation | AAC,GAC | N,D 215 | XP_016868866.1 |
MIR6848 - microRNA 6848 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |