Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGAAGCTGTTCCTGTGGCTCCA[A/G]AAGATCTTCTTTCTAAAACAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THAP1 - THAP domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018105.2 | 517 | Silent Mutation | CTG,TTG | L,L 94 | NP_060575.1 | |
NM_199003.1 | 517 | Silent Mutation | TTC,TTT | F,F 28 | NP_945354.1 |