Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGAAGATTTCTCTTTTTTATTC[C/T]GGGGAGAGTCTGCTACATTTGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605190 | ||||||||||||||||||||
Literature Links: |
TRAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAM1 - translocation associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317804.1 | 1576 | Missense Mutation | CAG,CGG | Q,R 336 | NP_001304733.1 | |
NM_001317805.1 | 1576 | Missense Mutation | CAG,CGG | Q,R 281 | NP_001304734.1 | |
NM_014294.5 | 1576 | Missense Mutation | CAG,CGG | Q,R 367 | NP_055109.1 | |
XM_017013266.1 | 1576 | Missense Mutation | CAG,CGG | Q,R 271 | XP_016868755.1 | |
XM_017013267.1 | 1576 | Missense Mutation | CAG,CGG | Q,R 271 | XP_016868756.1 |