Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCTACCTGATTCATCATTGAA[A/G]GAGGTAAAACTGATCAGCATTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608413 | ||||||||||||||||||||
Literature Links: |
UBR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBR5 - ubiquitin protein ligase E3 component n-recognin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282873.1 | 8513 | Silent Mutation | TCC,TCT | S,S 2694 | NP_001269802.1 | |
NM_015902.5 | 8513 | Silent Mutation | TCC,TCT | S,S 2695 | NP_056986.2 | |
XM_005250962.4 | 8513 | Silent Mutation | TCC,TCT | S,S 1707 | XP_005251019.1 | |
XM_011517104.2 | 8513 | Silent Mutation | TCC,TCT | S,S 2682 | XP_011515406.2 | |
XM_011517105.2 | 8513 | Silent Mutation | TCC,TCT | S,S 2681 | XP_011515407.2 | |
XM_011517106.2 | 8513 | Silent Mutation | TCC,TCT | S,S 2676 | XP_011515408.2 | |
XM_017013533.1 | 8513 | Silent Mutation | TCC,TCT | S,S 2680 | XP_016869022.1 | |
XM_017013534.1 | 8513 | Silent Mutation | TCC,TCT | S,S 2679 | XP_016869023.1 |
UBR5-AS1 - UBR5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |