Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTCCTTAGTGCAATACTTTGGT[G/T]ACAACTTGGGGCGAAAAGTTAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604598 | ||||||||||||||||||||
Literature Links: |
OSGIN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSGIN2 - oxidative stress induced growth inhibitor family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126111.2 | 366 | Missense Mutation | GAC,TAC | D,Y 36 | NP_001119583.1 | |
NM_004337.2 | 366 | UTR 5 | NP_004328.1 | |||
XM_011517287.2 | 366 | UTR 5 | XP_011515589.1 | |||
XM_011517288.2 | 366 | Intron | XP_011515590.1 |