Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTGTCCATCTGCTGTGAGGCTG[C/G]AGCCATGAACAAAGGCTGGACCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF251 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF251 - zinc finger protein 251 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138367.1 | 1842 | Missense Mutation | TCC,TGC | S,C 529 | NP_612376.1 | |
XM_005272356.4 | 1842 | Intron | XP_005272413.1 | |||
XM_006716683.2 | 1842 | Intron | XP_006716746.1 | |||
XM_011517361.1 | 1842 | Intron | XP_011515663.1 |