Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTCCTGCTGCCTGTACTTTGG[C/T]TCATTGTTCAAACTCAAGGTACGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606393 | ||||||||||||||||||||
Literature Links: |
ADAMDEC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMDEC1 - ADAM like decysin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145271.1 | 290 | UTR 5 | NP_001138743.1 | |||
NM_001145272.1 | 290 | UTR 5 | NP_001138744.1 | |||
NM_014479.3 | 290 | Missense Mutation | CTC,TTC | L,F 24 | NP_055294.1 |
LOC101929294 - uncharacterized LOC101929294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |