Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTTTAAAGGTCACATTGGCTG[C/G]ATTCACCAATGTTTTTAGTTTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605599 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375839 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375839 - uncharacterized LOC105375839 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYPLA1 - lysophospholipase I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279356.1 | 577 | Intron | NP_001266285.1 | |||
NM_001279357.1 | 577 | Intron | NP_001266286.1 | |||
NM_001279358.1 | 577 | Intron | NP_001266287.1 | |||
NM_001279359.1 | 577 | Intron | NP_001266288.1 | |||
NM_001279360.1 | 577 | Intron | NP_001266289.1 | |||
NM_006330.3 | 577 | Intron | NP_006321.1 | |||
XM_005251127.4 | 577 | Missense Mutation | CCA,GCA | P,A 146 | XP_005251184.1 | |
XM_017012956.1 | 577 | Intron | XP_016868445.1 | |||
XM_017012957.1 | 577 | Intron | XP_016868446.1 | |||
XM_017012958.1 | 577 | Intron | XP_016868447.1 | |||
XM_017012959.1 | 577 | Intron | XP_016868448.1 | |||
XM_017012960.1 | 577 | Intron | XP_016868449.1 |