Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCAGGGTATCAGAAGGTGATGT[A/C]ATCCTCGTACAGGGACAGCAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606027 MIM: 607059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPSF1 - cleavage and polyadenylation specific factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A4 - solute carrier family 39 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280557.1 | 1862 | Missense Mutation | GAC,TAC | D,Y 146 | NP_001267486.1 | |
NM_017767.2 | 1862 | Missense Mutation | GAC,TAC | D,Y 619 | NP_060237.2 | |
NM_130849.3 | 1862 | Missense Mutation | GAC,TAC | D,Y 644 | NP_570901.2 | |
XM_006716599.1 | 1862 | Missense Mutation | GAC,TAC | D,Y 593 | XP_006716662.1 |