Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGAAGCTGGTGCCTCTCCAAGT[A/G]TTAATTTGTTCTTACAAATGTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608479 | ||||||||||||||||||||
Literature Links: |
LRRC69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC69 - leucine rich repeat containing 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129890.1 | 94 | Silent Mutation | GTA,GTG | V,V 326 | NP_001123362.1 |
SLC26A7 - solute carrier family 26 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282356.1 | 94 | UTR 5 | NP_001269285.1 | |||
NM_001282357.1 | 94 | UTR 5 | NP_001269286.1 | |||
NM_052832.3 | 94 | Intron | NP_439897.1 | |||
NM_134266.1 | 94 | Intron | NP_599028.1 |