Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAATCTCAGAAGGACTGTGCAA[A/G]TCAATGAGTCGCTTGTGAATTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375847 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375847 - uncharacterized LOC105375847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS20 - ribosomal protein S20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001023.3 | 468 | Silent Mutation | GAC,GAT | D,D 90 | NP_001014.1 | |
NM_001146227.1 | 468 | Silent Mutation | GAC,GAT | D,D 90 | NP_001139699.1 |
SNORD54 - small nucleolar RNA, C/D box 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |