Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCATGGATCTTTGGCTTGCGGG[C/G]GTTGAAAACGTGTCATATTTTTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DEFB135 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEFB135 - defensin beta 135 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DEFB136 - defensin beta 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033018.2 | 206 | Missense Mutation | CCC,CGC | P,R 69 | NP_001028190.2 |