Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGTCCAGGGCGAGCCTCGCTTC[A/G]GCCCCAGCCTCTGCCGCCTCCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC642574 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC642574 - IQ motif and ankyrin repeat domain-containing protein LOC642574 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRIB - scribbled planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015356.4 | 4243 | Silent Mutation | GCC,GCT | A,A 1412 | NP_056171.3 | |
NM_182706.4 | 4243 | Silent Mutation | GCC,GCT | A,A 1412 | NP_874365.3 |