Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTCCATCCTGGCAGGCGGCTGT[A/G]GTGGTTTGAAGAGTTTGGACAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603912 | ||||||||||||||||||||
Literature Links: |
EIF3H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF3H - eukaryotic translation initiation factor 3 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003756.2 | 949 | Missense Mutation | CCA,CTA | P,L 308 | NP_003747.1 |
LOC105375713 - uncharacterized LOC105375713 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |