Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTTGAAGAAGGACAAAAAGGA[A/G]TAGGCGACGGTACAGTTAGCTGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603001 | ||||||||||||||||||||
Literature Links: |
UBE2V2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2V2 - ubiquitin conjugating enzyme E2 V2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003350.2 | 156 | Missense Mutation | ATA,GTA | I,V 26 | NP_003341.1 | |
XM_005251300.2 | 156 | UTR 5 | XP_005251357.1 | |||
XM_011517583.2 | 156 | Missense Mutation | ATA,GTA | I,V 54 | XP_011515885.1 | |
XM_017013808.1 | 156 | Missense Mutation | ATA,GTA | I,V 23 | XP_016869297.1 |