Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACCAGCACAATCTGTACCGTCG[C/T]CGCCGCTCCCGTGGTCCCAGCGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603303 | ||||||||||||||||||||
Literature Links: |
TNKS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNKS - tankyrase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003747.2 | 1010 | Missense Mutation | GCC,GTC | A,V 100 | NP_003738.2 | |
XM_006716263.3 | 1010 | Missense Mutation | GCC,GTC | A,V 100 | XP_006716326.1 | |
XM_011543845.2 | 1010 | Missense Mutation | GCC,GTC | A,V 100 | XP_011542147.1 | |
XM_011543846.2 | 1010 | Missense Mutation | GCC,GTC | A,V 100 | XP_011542148.1 | |
XM_011543847.2 | 1010 | Missense Mutation | GCC,GTC | A,V 100 | XP_011542149.1 |