Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGATCACGGCCCGCAGCCTCTA[C/T]CCGGCAGACTTTAACATCCAGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611353 | ||||||||||||||||||||
Literature Links: |
INTS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTS10 - integrator complex subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018142.2 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | NP_060612.2 | |
XM_005273555.3 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_005273612.1 | |
XM_005273556.3 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_005273613.1 | |
XM_005273557.3 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_005273614.1 | |
XM_006716355.2 | 62 | UTR 5 | XP_006716418.1 | |||
XM_017013606.1 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_016869095.1 | |
XM_017013607.1 | 62 | Silent Mutation | TAC,TAT | Y,Y 36 | XP_016869096.1 | |
XM_017013608.1 | 62 | UTR 5 | XP_016869097.1 | |||
XM_017013609.1 | 62 | UTR 5 | XP_016869098.1 |