Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTGTTGTCAAAGTCTGCCTTC[A/G]TTGCTCCCATCTTCACTGCAACAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138300 | ||||||||||||||||||||
Literature Links: |
GSR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSR - glutathione-disulfide reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000637.3 | 1387 | Missense Mutation | ACG,ATG | T,M 500 | NP_000628.2 | |
NM_001195102.1 | 1387 | Missense Mutation | ACG,ATG | T,M 471 | NP_001182031.1 | |
NM_001195103.1 | 1387 | Missense Mutation | ACG,ATG | T,M 447 | NP_001182032.1 | |
NM_001195104.1 | 1387 | Missense Mutation | ACG,ATG | T,M 418 | NP_001182033.1 |