Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCTGGCGCAGGTGCAGCACCC[A/C]CTGCTGCATTACCTCAGCGTCCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615791 | ||||||||||||||||||||
Literature Links: |
FAM160B2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM160B2 - family with sequence similarity 160 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022749.5 | 417 | Silent Mutation | CCA,CCC | P,P 122 | NP_073586.5 | |
XM_005273602.3 | 417 | Silent Mutation | CCA,CCC | P,P 122 | XP_005273659.1 | |
XM_006716381.2 | 417 | Silent Mutation | CCA,CCC | P,P 122 | XP_006716444.1 | |
XM_006716385.3 | 417 | Silent Mutation | CCA,CCC | P,P 19 | XP_006716448.1 | |
XM_017013732.1 | 417 | Silent Mutation | CCA,CCC | P,P 122 | XP_016869221.1 | |
XM_017013733.1 | 417 | Silent Mutation | CCA,CCC | P,P 64 | XP_016869222.1 | |
XM_017013734.1 | 417 | Silent Mutation | CCA,CCC | P,P 64 | XP_016869223.1 |
NUDT18 - nudix hydrolase 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |