Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGCATCTCTGGGAGTAAAAAT[G/C]CAAAATATCCAGTGTCACGTAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609562 | ||||||||||||||||||||
Literature Links: |
CPA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPA6 - carboxypeptidase A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020361.4 | 1473 | Missense Mutation | GCA,GGA | A,G 406 | NP_065094.3 | |
XM_011517570.2 | 1473 | Missense Mutation | GCA,GGA | A,G 258 | XP_011515872.1 | |
XM_017013646.1 | 1473 | Missense Mutation | GCA,GGA | A,G 258 | XP_016869135.1 | |
XM_017013647.1 | 1473 | Intron | XP_016869136.1 |
LOC102724708 - uncharacterized LOC102724708 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |